MSA™ Ultra-Twin™ Respirators
Turned-in edge for a tight seal against the face and large chin cup for easy positioning
Manufacturer: MSA™ 480263
Requires : Comfo Series cartridges and filters (see separate listings).
Description
- Five suspension headstraps and roller buckles secure the facepiece while minimizing hair entanglement
- Wide polycarbonate lens is coated to resist scratching and chemical attack
- Available in Hycar™ rubber and silicone
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Silicone | |
Black | |
Small | |
Nosecup and speaking diaphragm |
NIOSH | |
Ultra-Twin™ Full Facepiece Respirator | |
Asbestos abatement, painting, sanding, grinding, welding | |
Polycarbonate |