MSA™ Ultra Elite™ Twin Cartridge Full-Facepiece Respirators
Designed to provide exceptional peripheral and downward vision.
Manufacturer:  MSA™ 10016756
 View more versions of this product
Catalog No. 19-065-242
Includes: Twin cartridge adapter
Requires : Comfo series aerosol filters and chemical cartridges
Description
Description
When used with appropriate filter cartridges, this NIOSH-approved facepiece lens/respirator combination provides an ideal defense against numerous hazards including particulates, dust, projectiles, and splashing liquids in toxic atmospheres not deemed Immediately Dangerous to Life or Health (non-IDLH).
  • Wide-vision lens
  • Black, SoftFeel™ silicone respirator
  • Large adjustment tabs to facilitate donning and doffing and a five-point rubber head harness
  • Standard speaking diaphragm and nose-cup
  • Suits a wide range of industrial applications including asbestos abatement, paper and pulp manufacturing and spray painting
Order Info
Additional accessories such as lens cover (Cat. No. 19-815-684) and speech diaphragm (Cat. No. 18-999-3861) available separately.
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
SoftFeel Silicone
Ultra Elite Facepiece Respirator
Asbestos abatement, painting, sanding, grinding, welding
Twin cartridge adapter
EN 136 class 3, RU: GOST R 12.4.189-99, KZ: ST RK, GOST R 12.4.189-99, BY: STB GOST R 12.4.189-2006, AS NZS 1716:2003
Medium
Filter cartridge
Black wide-vision