MSA™ Toxgard II Gas Monitor: Sensor Kit, O2
This is a kit containing the MSA 10019727 oxygen sensor. These sensors are typically used in oxygen deficiency air monitoring, are normally exposed to ambient conditions, and are not operated below 2% oxygen concentration for extended time periods.
Manufacturer:  MSA™ 813723
Catalog No. 29-636-6420
Description
Description
These sensors also compensate for changes in barometric pressure and can be used to monitor for oxygen purity.
Specifications
Specifications
Sensor Kit, Oxygen
0 to 25%
Oxygen