MSA™ Tee Blocks and Pigtails
Supplier:  MSA™ 68850
 View more versions of this product
Catalog No. 2985036
Description
Description
Used to connect a bank of cylinders together
  • Coupler tees make a connection between the supply cylinder valve and the pigtail
  • Outlet connection are constructed of brass, while the pigtails joining the supply cylinders are made of tubular copper
  • 27 in. long air pigtail
Specifications
Specifications
Air tee block
Connection between the supply cylinder valve and the pigtail and the outlet connection
Compressed airline respirator