Description
Used to connect a bank of cylinders together
- Coupler tees make a connection between the supply cylinder valve and the pigtail
- Outlet connection are constructed of brass, while the pigtails joining the supply cylinders are made of tubular copper
- 27 in. long air pigtail
Specifications
Air pigtail | |
68.58 cm | |
Joining the supply cylinders | |
Tubular Copper |
27 in. | |
3000 psi | |
Compressed airline respirator | |
Pigtail |