missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MSA™ TechnaCurv™ Full-Body Vest-Style Harnesses

Made of soft, flexible webbing

$167.69 - $330.98

Specifications

Certifications/Compliance OSHA, ANSI Z359.1 and A10.32, CSA Z259.10. Certified according to EN 361and EN 358
Color Red
For Use With (Application) Confined space, construction
Product Type Full-body harness
Style Vest,Vest
View More Specs

Products 9
Catalog Number Mfr. No. Buckle Style (Leg) D-Ring Location Includes Size Price Quantity  
Catalog Number Mfr. No. Buckle Style (Leg) D-Ring Location Includes Size Price Quantity  
19-121-499
View Documents
MSA™
10041600
Tongue Back Shoulder padding, Qwik-fit chest strap, manual X-Large
Each for $230.75
 
19-130-3891
View Documents
MSA™
10041591
Tongue Back Qwik-fit chest strap, manual Standard
Each for $182.14
 
17-200-103
View Documents
MSA™
10041592
Tongue Back Qwik-fit chest strap, manual X-Large
Each for $167.69
 
19-160-0187
View Documents
MSA™
10041599
Tongue Back Shoulder padding, Qwik-fit chest strap, manual Standard
Each for $245.91
 
17-200-104
View Documents
MSA™
10041595
Tongue Back, Hips Qwik-fit chest strap, manual Standard
Each for $170.66
 
17-200-107
View Documents
MSA™
10041607
Tongue Back, Chest, Hips Qwik-fit chest strap, manual Standard
Each for $207.59
 
17-200-108
View Documents
MSA™
10041608
Tongue Back, Chest, Hips Qwik-fit chest strap, manual Standard
Each for $200.71
 
17-200-112
View Documents
MSA™
10041616
Secure-Fit Back, Chest, Hips Shoulder and leg padding, Secure-Fit leg straps, Qwik-fit chest strap, manual X-Large
Each for $330.98
 
17-200-111
View Documents
MSA™
10041615
Secure-Fit Back, Chest, Hips Shoulder and leg padding, Secure-Fit leg straps, Qwik-fit chest strap, manual Standard
Each for $325.20
 
Description

Description

Optional Secure-Fit leg buckles with positive engagement windows allow you to see buckle is properly engaged—no need to listen for a “click” on a noisy site.

  • Soft, flexible curvilinear torso webbing moves with the wearer for improved comfort and a secure fit
  • Webbing curves to fit around the neck, so it will not chafe
  • Lightweight polymer Y-back D-locator pad allows complete adjustment of back D-ring for proper fit
  • Durable visco-elastic shoulder and sub-pelvic pads permit air to circulate; keep wearer cool and comfortable
  • Single-hand operation for easy donning and doffing
  • Feature a standard Qwik-Fit™ chest strap

Order Info

Maximum Capacity: 400 lb. (181kg).

Compliance

Meets applicable OSHA standards; ANSI Z359.11-2014: CSA Z259.10-12 requirements.

Specifications

Specifications

OSHA, ANSI Z359.1 and A10.32, CSA Z259.10. Certified according to EN 361and EN 358
Confined space, construction
Vest,Vest
Red
Full-body harness
Teflon-coated Polyester
SDS