MSA™ TechnaCurv™ Full-Body Vest-Style Harnesses
Made of soft, flexible webbing
Manufacturer:  MSA™ 10041600
 View more versions of this product
Catalog No. 19-121-499
Description
Description
Optional Secure-Fit leg buckles with positive engagement windows allow you to see buckle is properly engaged—no need to listen for a “click” on a noisy site.
  • Soft, flexible curvilinear torso webbing moves with the wearer for improved comfort and a secure fit
  • Webbing curves to fit around the neck, so it will not chafe
  • Lightweight polymer Y-back D-locator pad allows complete adjustment of back D-ring for proper fit
  • Durable visco-elastic shoulder and sub-pelvic pads permit air to circulate; keep wearer cool and comfortable
  • Single-hand operation for easy donning and doffing
  • Feature a standard Qwik-Fit™ chest strap
Order Info
Maximum Capacity: 400 lb. (181kg).
Compliance
Meets applicable OSHA standards; ANSI™ Z359.11-2014: CSA Z259.10-12 requirements.
Specifications
Specifications
Tongue
400 lb.
Back
Shoulder padding, Qwik-fit chest strap, manual
Vest
Red
Tongue
OSHA, ANSI Z359.1 and A10.32, CSA Z259.10. Certified according to EN 361and EN 358
Confined space, construction
X-Large
Teflon-coated Polyester
Full-body harness