MSA™ Ultra Elite Spectacle Kit for Self-Contained Breathing Apparatus (SCBA)
Manufacturer:  MSA™ 493581
 View more versions of this product
Catalog No. 29811613
Includes: Note: These do not include lenses - just wire support and frames.
Description
Description
Kits allow Ultra Elite™ facepiece to accommodate corrective lenses. Include wire support and rubber guide for easy custom adjustment and S-7 shaped metal spectacles frame for use with 44mm universal bridge corrective lenses. Choose from either side-wire or center support kit.
Specifications
Specifications
Wire support, rubber guide, pair of S-7 shaped metal frame spectacles
Msa™ respirator utilizing ultra-elite facepiece, spectacle insert kit
Center Wire Support