missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MSA™ Sirius™ 4-Gas Multigas Detector - Deluxe Model

Combines PID functionality with a four-gas monitor

$6,359.46 - $6,359.46

Specifications

Accuracy ±10%
Certifications/Compliance 47CFR, UL913, CSA 22.2
Depth (English) 2.62 in.
Depth (Metric) 6.65cm
For Use With (Application) Leak detection, perimeter monitor, spill delineation, remediation, arson investigation, diesel fuel TVL monitoring, decontamination, PPE determination
View More Specs

Includes: All monitors come with a 10.6 eV lamp and standard lamp cap, lithium battery, 10 ft. polyurethane sampling line, 1 ft. probe, retractable carrying line with belt clip, calibration kit, and plastic carrying case. The CH4 model also includes a lamp cleaning kit.

Products 1
Catalog Number Mfr. No. Sensors Model Price Quantity  
Catalog Number Mfr. No. Sensors Model Price Quantity  
19-123-398
View Documents
MSA™
ASIRIUSAL111A0111D101SC
LEL, O2, CO, H2S LEL Kit, without PID Datalog
Each for $6,359.46
 
Description

Description

Integrates high-performance photoionization detector sensor with a multigas detector for up to four gases. Users are able to simultaneously monitor for volatile organic compounds (VOCs) with low vapor pressures while measuring for combustible, toxic, and oxygen-deficient atmospheres within one reliable, easy-to-use, durable unit.

  • Measures VOC readings first for greater accuracy of ±10% or 2ppm over full range
  • Best in class humidity performance
  • Notifies failure or removal of lamp or ion chamber
  • Features an overrange indicator, external exhaust, and multidirectional audio and visual alarms
  • Lamps are easy to replace and clean

For use in:

  • Leak detection
  • Perimeter monitor
  • Spill delineation
  • Remediation
  • Arson investigation
  • Diesel fuel TLV monitoring (Marine Chemist)
  • Decontamination
  • PPE Determination

Compliance

Meets 47CFR part 15. UL913 6th Edition and CSA 22.2 No. 157 for use in hazardous locations Class I, Divisions I, Groups A, B, C, and D. Temperature class T3C. Meets Canadian combustible performance standard CSA 22.2 No. 152 for methane only.

Specifications

Specifications

±10%
2.62 in.
Leak detection, perimeter monitor, spill delineation, remediation, arson investigation, diesel fuel TVL monitoring, decontamination, PPE determination
6.5 in.
Monitor, 10.6 eV lamp and standard lamp cap, lithium battery, 10 ft. polyurethane sampling line, 1 ft. probe, retractable carrying line with belt clip, calibration kit and plastic carrying case
0.1% Vol O2, 1ppm H2S, 1ppm CO, 1% LEL
9.19cm
47CFR, UL913, CSA 22.2
6.65cm
Multigas Detector
16.51cm
0 to 25% O2, 0 to 500ppm CO, 0 to 200ppm H2S, 0 to 100% LEL
3.62 in.
SDS
Product Certifications