MSA™ SCBA Replacement Cylinders: With Valve
Manufacturer:  MSA™ 807587
 View more versions of this product
Catalog No. 29811891
Specifications
Specifications
Firefighting
Carbon Wrapped Aluminum
1.27cm3
Valve
3.3kg
45 cu. ft., 1.27cm3
SCBA
Gas Cylinder, H-30 Stealth
45 cu. ft.
4500psi
7 lb.
30 min.
Documents