MSA™ Sampling Line for Gas Detection
For use with combustible gas indicator
Manufacturer:  MSA™ 11354
Catalog No. 2985183
Description
Description
  • Made of rubber
  • Length: 5 ft. (152.4cm)
  • Diameter: 0.25 in. (6.3mm)
Specifications
Specifications
Sampling Line