MSA™ Safe Escape CBRN Training Hoods
Manufacturer:  MSA™ 10066785
 View more versions of this product
Catalog No. 29-635-6395
Description
Description
Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
Certifications
NIOSH - 10052317
Specifications
Specifications
NIOSH
CBRN incidents, civil disturbances, clandestine labs
Training
Training Hood
Emergency escape respirator
Medium