Description
Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
Certifications
NIOSH - 10052317
Specifications
NIOSH | |
CBRN incidents, civil disturbances, clandestine labs | |
Training |
Training Hood | |
Emergency escape respirator | |
Small |