MSA™ Safe Escape CBRN Respirators
Designed to escape sudden chemical, biological, radiological, or nuclear hazards
Manufacturer: MSA™ 10066791
Description
- Translucent hood with one-piece large bonded lens offers secure protection and a wide field of view
- Canister is permanently bonded to the respirator for a secure connection
- Rubber neck seal provides a comfortable and reliable seal
- Elastic hair tie allows for the securing of hair before donning the respirator
- Nose cup helps reduce lens fogging and carbon dioxide buildup
- Hexagonal storage case is strong, lightweight, and easily accessible
- 15 minute air supply
Standard
- Color-coded labels on respirator
- Black/gray labels on respirator
Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
Certifications
NIOSH - 10052317
Specifications
NIOSH | |
15 min. | |
Bonded lens and cbrn canister, high stretch rubber neck seal, nose cup | |
Medium |
Standard Escape Respirator | |
CBRN incidents, civil disturbances, clandestine labs | |
Standard |