MSA™ Safe Escape CBRN Respirators
Designed to escape sudden chemical, biological, radiological, or nuclear hazards
Manufacturer:  MSA™ 10066787
 View more versions of this product
Catalog No. 29-635-6391
Description
Description
  • Translucent hood with one-piece large bonded lens offers secure protection and a wide field of view
  • Canister is permanently bonded to the respirator for a secure connection
  • Rubber neck seal provides a comfortable and reliable seal
  • Elastic hair tie allows for the securing of hair before donning the respirator
  • Nose cup helps reduce lens fogging and carbon dioxide buildup
  • Hexagonal storage case is strong, lightweight, and easily accessible
  • 15 minute air supply
Standard
  • Color-coded labels on respirator
  • Tactical
    • Black/gray labels on respirator
    Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
    Certifications
    NIOSH - 10052317
    Specifications
    Specifications
    NIOSH
    15 min.
    Bonded lens and cbrn canister, high stretch rubber neck seal, nose cup
    Small
    Tactical Escape Respirator
    CBRN incidents, civil disturbances, clandestine labs
    Tactical