MSA™ Replacement Suspensions for V-Gard™ Caps and Hats
Designed for easy clip-in to the V Gard hat or cap shell, users can adjust the liner for a secure and comfortable fit.
Manufacturer:  MSA™ 10087218
 View more versions of this product
Catalog No. 19-049344
Description
Description
Staz-On Suspension
Comfortable four-point webbed suspension features adjustable dual nylon crown straps, a perforated vinyl sweatband with pinlock closure
Fas-Trac Ratchet Suspension
Four-point webbed suspension features adjustable dual nylon crown straps, a perforated vinyl sweatband, and a large easy-adjustment knob closure
1-Touch Suspension
One-handed slide adjustment; easy squeeze opening
Specifications
Specifications
4-Point webbed suspension, adjustable dual Nylon crown straps, perforated Vinyl sweatband
Headband, nape strap, sweatband and adjustable crown straps
Staz-On™ Pin Lock Suspension System
16.51 to 20.32 cm
V-Gard™ Hard Hat and Cap
Plastic, Nylon
Standard
6.5 to 8 in.