Please login to your online account to display your discounted pricing

MSA™ Replacement Facepiece Assemblies for Ultra Elite™ and Ultravue™ SCBAs

$773.97 - $776.94

Specifications

Certifications NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002
Color Black
For Use With (Application) For monitoring air cylinder contents, firefighting
View More Specs
Products
Catalog Number Mfr. No. Includes Material Size For Use With (Equipment) Price Quantity    

189993162

 
msa™
815854
Quarter turn quick-connect adapter, rubber head harness and nosecup Hycar™ Rubber Medium Ultralite mmr and custom 4500 mmr air mask Each for $773.97

19-005-402B

 
msa™
10024411
Speed-on™ head harness, nosecup Hycar™ Rubber Medium Firehawk™ mmr air mask Each for $773.97

19-005-402C

 
msa™
10024431
Speed-on™ head harness, nosecup Hycar™ Rubber Large Firehawk™ mmr air mask Each for $776.94
Description & Specifications

Specifications

Certifications NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002
Color Black
For Use With (Application) For monitoring air cylinder contents, firefighting