Please login to your online account to display your discounted pricing

MSA™ Replacement Facepiece Assemblies for AirHawk™ MMR

$661.35 - $661.35

Specifications

Certifications NIOSH
Color Black
For Use With (Equipment) Airhawk™ mmr air mask
Product Type Pressure Demand Facepiece
For Use With (Application) Chemical and petrochemical, hazardous materials, confined space entry, firefighting operation
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-005-406A

 
msa™
10037650
Small Each for $661.35

19-005-406C

 
msa™
10037652
Large Each for $661.35
Description & Specifications

Specifications

Certifications NIOSH
Color Black
For Use With (Equipment) Airhawk™ mmr air mask
Product Type Pressure Demand Facepiece
For Use With (Application) Chemical and petrochemical, hazardous materials, confined space entry, firefighting operation
Harness Style Rubber
Includes Head harness and nosecup
Material Hycar™ Rubber
Material (Faceshield) Hycar™