missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MSA™ Orion™ Multigas Detector: Sample Probes

Manufacturer:  MSA™ 710465

 View more versions of this product

Catalog No. 2985143



Description

Description

Specifications

Specifications

9 x 7 x 5 in. (22.8 x 17.7 x 12.7cm)
5 in.
9 in.
Polyurethane Coil
17.7 cm
Sampling Line Probe
12.7 cm
22.8 cm
7 in.
SDS
Documents
Product Certifications
Promotions

Promotions