MSA™ OptimAir™ 6A PAPR with OptiFilter™ Cartridges
Ideal for asbestos and lead paint abatement, hazardous materials cleanup, chemical processing, and similar applications
Manufacturer: Msa Canada 494236
Includes: Either the hood or the full facepiece plus breathing tube, battery charger, belt, and motor/blower module. Cat. No. 18-999-3110 also includes two OptiFilter XL filter cartridges.
Description
Protects against particulates, toxic gases and vapors, or a combination of these hazards. Filters contaminants from the outside air and provides constant airflow. Sold as a complete assembly.
For homeland security applications, a version with Tychem™ SL hood and CBA/RCA canisters is also available. (Call for details.)
Facepiece/Hood Option
- Available with comfortable, wide-vision Ultravue™ Hycar™ rubber full-facepiece or with a Tyvek™ hood
- Efficient, long-lasting motor/blower assembly is belt-mounted and has a threaded connector for quick attachment of the filter cartridges
Order Info
Replacement parts are available.
Compliance
Approved by NIOSH and MSHA.
Specifications
NIOSH MSHA | |
Black hycar™ ultravue™ facepiece, motor/blower module, nicad battery, facepiece, breathing tube, belt and battery charger | |
Air Purifying Respirator | |
Medium |