MSA™ OptiFilter™ Cartridges for OptimAir™ Respirators
Line of cartridges and filters is compatible with OptimAir 6A and OptimAir MM 2K PAPR powered air-purifying respirators
$227.62 - $227.62
Specifications
Certifications/Compliance NIOSH
Filter Efficiency 99.97%
Quantity 6/Pk.
For Use With (Equipment) Optimair™ 6a and optimair™ mm 2k powered air purifying respirator powered air purifying respirator
View More Specs
Products 1
Catalog Number Mfr. No. Cartridge Type Price Quantity  
Catalog Number Mfr. No. Cartridge Type Price Quantity  
189993427
View Documents
MSA™
496081
OptiFilter™ XL type HE Cartridge
Pack of 6 Each for $227.62
Description
Description
  • Type HE OptiFilter XL cartridge is a HEPA filter with a 99.97% filter efficiency level for protection against all particulate aerosols; CBA/RCA model suits Homeland Security applications GMA cartridge is approved for not more than 1000ppm organic vapors; particulate filter (99.97% filter efficiency level) is effective against all particulate aerosols GMC cartridge is approved for not more than 1000ppm organic vapors; chloride, chlorine dioxide, hydrogen chloride, sulfur dioxide, and escape only from hydrogen sulfide; particulate filter (99.97% filter efficiency level) is effective against all particulate aerosols
Specifications
Specifications
NIOSH
99.97%
6/Pk.
Optimair™ 6a and optimair™ mm 2k powered air purifying respirator powered air purifying respirator