missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MSA™ Nosecups

$156.69 - $158.83

Specifications

Color Black
For Use With (Application) To reduce lens fogging
Product Type Nosecup
Material Rubber
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18-999-3654
View Documents Promotion Details
MSA™
813140
Large
Each for $156.69
 
19-037-292
View Documents Promotion Details
MSA™
813139
Medium
Each for $158.83
 
Description

Description

Specifications

Specifications

Black
To reduce lens fogging
Nosecup
Rubber
SDS
Product Certifications