MSA™ NFL™ V-Gard™ Caps
Official NFL insignias help promote regular use on the job
Manufacturer: MSA™ 818384
Description
- MSA V-Gard caps are constructed of high-density polyethylene
Compliance
Third-party certified to meet the Type I, Class E requirements of ANSI/ISEA Z89.1 and CSA Z94.1.
NFL Properties, the NFL and its member clubs have not produced or endorsed this headgear, make no claims about its effectiveness, and will have no liability for any claims related to such headgear.
Specifications
Meets or exceeds ANSI Z89.1-2003; Type I; Classes G and E. | |
Arizona Cardinals | |
HDPE | |
1-Touch |