MSA™ NFL™ Logo V-Gard™ Caps - AFC Teams
Official NFL insignias help promote regular use on the job
Manufacturer:  MSA™ 818393
 View more versions of this product
Catalog No. 19-0881093
Description
Description
  • MSA™V-Gard™ one-piece caps Constructed of high-density polyethylene
Compliance
Meet or exceed all applicable requirements of ANSI Z89.1-2003 for Type I helmets, Classes G and E.
NFL Properties, the NFL and its member clubs have not produced or endorsed this headgear, make no claims about its effectiveness, and will have no liability for any claims related to such headgear.
Specifications
Specifications
Meets or exceeds ANSI Z89.1-2003; Type I; Classes G and E.
Denver Broncos
1-Touch