MSA™ Model RP Combination Regulators
A fixed flow design provides constant and accurate flow to the gas detector simplifying the test and calibration process and saving calibration gas
Manufacturer: MSA™ 711174
Description
- Two regulators in one, designed for use with all Model RP and Econo-Cal calibration cylinders
- No ON/OFF valve is necessary, because the regulator automatically turns itself off when not in use
- Trigger activation allows a bump test or calibration check in the "SQUEEZE" mode or fixed-flow calibration in the up or "LOCKED" position
Specifications
Respirator Assembly | |
1.5L/min. | |
Calibration tubing |
Model RP | |
Agriculture, chemical, construction, electric utility, fire fighting, with calibration tubing |