missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MSA™ Model RP Calibration Check Kits
Quick and convenient method for checking the response of gas detection instruments
$534.66 - $1,013.01
Catalog Number | Mfr. No. | Type | Flow Rate | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Flow Rate | Price | Quantity | ||||
189993342
|
MSA™
467895 |
Replacement Regulator | 0.25 L/min. |
Each for $534.66
|
|
18-999-3236
|
MSA™
477149 |
Calibration Kit | 0.25 L/min. |
Each for $1,013.01
|
|
Description
Kits available with 0.25L/min. or 1.5L/min. regulators.
Order Info
Cylinders are sold separately. (Measure approximately 14.25L × 3.5 in. diameter.)
Certifications
All calibration accessories are certified and tested for accuracy.