MSA™ FP5K™ Tie-Back Shock Absorbing Lanyards
Unique carabiner offers 2268kg (5000 lb.) gate strength in any direction
Manufacturer:  MSA™ 10047084
 View more versions of this product
Catalog No. 19-130-3879
Description
Description
  • Carabiner can be tied back directly to the lanyard and enables maximum inside clearance with a 2.54cm (1 in.) gate opening
  • Carabiner body shape helps avoid confusion with standard connectors not designed for tie-back applications
  • Monster-Edge™ Webbing has a 5443kg (12,000 lb.) minimum breaking strength and designed for increased abrasion resistance
  • Internal wear indicator alerts user when lanyard should no longer be used due to severe abrasion
  • Keeps fall arrest forces below 900 lb./ft
  • 1.82m (6 in.) adjustable
Certifications
CSA certified
Compliance
Meets applicable OSHA standards; ANSI™ A10.32, ANSI Z359.1, and ANSI Z359.13; CSA Z259.11-2005 requirements. CSA certified.
Specifications
Specifications
FP5K Carabiner
6 in.
Nylon webbing
Single-Leg
1.82 m