missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MSA™ FP Diamond™ Energy-Absorbing Lanyard
Lightweight and streamlined design
Manufacturer: MSA™ 10045936
Description
Connects the back (dorsal) D-ring of MSA full-body harnesses to a qualified anchorage and anchorage connector
- Keeps average fall arrest forces below 900 lb.
- Durable polyester sheath for increased abrasion and UV resistance
- Pouchless design for lighter weight and more comfortable wear
- Aluminum or zinc-plated steel snaphooks with auto-close and lock
- Highly visible deployment tag
- Swivel HL200 harness connection
- Single-leg configuration is bi-directional to make training easier, while twin-leg configuration allows for 100% tie off
Compliance
- US: ANSI/ASSE Z359.13, Z359.3; OSHA 29 CFR 1910.66, ASTM F887-005
- CAN: Z259.11-05
Specifications
HL2000 | |
Single-Leg | |
6 ft. | |
Polyester |