missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MSA™ FP Diamond™ Energy-Absorbing Lanyard

Lightweight and streamlined design

$85.76 - $85.76

Specifications

Length (English) 6 ft.
Material Polyester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Connector Types Leg Configuration Price Quantity  
Catalog Number Mfr. No. Connector Types Leg Configuration Price Quantity  
19-130-3894 MSA™
10045936
HL2000 Single-Leg Each for $85.76
Specifications

Description

Connects the back (dorsal) D-ring of MSA full-body harnesses to a qualified anchorage and anchorage connector

  • Keeps average fall arrest forces below 900 lb.
  • Durable polyester sheath for increased abrasion and UV resistance
  • Pouchless design for lighter weight and more comfortable wear
  • Aluminum or zinc-plated steel snaphooks with auto-close and lock
  • Highly visible deployment tag
  • Swivel HL200 harness connection
  • Single-leg configuration is bi-directional to make training easier, while twin-leg configuration allows for 100% tie off

Compliance

  • US: ANSI/ASSE Z359.13, Z359.3; OSHA 29 CFR 1910.66, ASTM F887-005
  • CAN: Z259.11-05

Description & Specifications

Specifications

6 ft.
Polyester