MSA™ Flexi-Filter™ Pads for Advantage™ Respirators
Supplier: MSA™ 818347
Specifications
EN 143, NIOSH, USA | |
Black | |
Advantage™ Flexi-Filters™ Pad | |
Asbestos abatement, painting, sanding, grinding, welding | |
Nuisance level OV removal |
Bayonet | |
N95 Filter | |
95% | |
Advantage™ respirator | |
2/Pk. |