MSA™ Flexi-Filter™ Pads for Advantage™ Respirators
Manufacturer:  MSA™ 818342
 View more versions of this product
Catalog No. 2984819
Description
Description
Specifications
Specifications
EN 143, NIOSH, USA
Magenta
Advantage™ Flexi-Filters™ Pad
Asbestos abatement, painting, sanding, grinding, welding
Filter pad only
Bayonet
P100 Filter
99.97%
Advantage™ respirator
2/Pk.