MSA™ Flexi-Filter™ Pads for Advantage™ Respirators
Manufacturer: MSA™ 818354
Specifications
EN 143, NIOSH, USA | |
Bayonet | |
P95 Filter | |
2/Pk. | |
Advantage™ respirator |
Black | |
95% | |
Filter pad only | |
Asbestos abatement, painting, sanding, grinding, welding | |
Advantage™ Flexi-Filters™ Pad |