MSA™ Flexi-Filter™ Pads for Advantage™ Respirators
Manufacturer:  MSA™ 818344
 View more versions of this product
Catalog No. 19-052-712
Description
Description
Specifications
Specifications
EN 143, NIOSH, USA
Bayonet
P100 Filter
2/Pk.
Advantage™ respirator
Magenta
99.97%
Nuisance level AG, HF removal
Asbestos abatement, painting, sanding, grinding, welding
Advantage™ Flexi-Filters™ Pad