MSA™ Evolution™ 5200 Thermal Imaging Camera
Delivers high-sense image quality up to 320°F, matching the temperature range at which most structural firefighting occurs
Manufacturer:  MSA™ 10063765
 View more versions of this product
Catalog No. 19-130-4003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Camera offers a super-high Instantaneous Scene Dynamic Range† (ISDR) of 4795, providing clearer, sharper, more detailed images over the entire temperature range. RFI shielded unit is situated inside a high-impact housing with external rubber bumpers and internal mounting foam (which insulates sensor and electronics).
Performance Features
  • Super-wide High Sense operating range allows camera to stay in high sense mode longer
  • High sensitivity in Low Sense mode provides better image quality at higher scene temperatures
  • Increased ambient temperature tolerance permits unit to stay in high heat 60-100% longer
  • Indigo 160 x 120 microbolometer offers excellent image quality in a small format thermal imager
  • 30Hz frame rate reduces the choppiness associated with lower frame rates
  • Optional “Heat Seeker PLUS” color detail allows user to easily locate hot spots and the seat of the fire
Additional Features
  • “Instant on” with instant mode switching; 15% Low Sense activation
  • Warnings on all shut downs
  • Shutter indicator
  • Optional Quick-Temp digital temperature display with temperature scale measurement also available
†Instantaneous Scene Dynamic Range is the combined value of a sensor's sensitivity and temperature range to determine the number of temperature differences that can be seen at any given instant.
‡Also includes reflective kit.
Specifications
Specifications
110/220V
Evolution™ 5200
67
Charger, Quick-Temp/Heat Seeker PLUS option, retractable lanyard, carrying case, two batteries, Reflective kit
Thermal imaging camera