missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MSA™ ESP™ II Voice Amplifier
Hands-free voice amplification for local, short-range communication
Manufacturer: MSA™ 10026265
Description
Battery-powered, self-contained communications unit for use with the Millennium™ and Advantage™ 1000 facepieces.
- Provides hands-free voice amplification for local, short-range communication
- Consists of a microphone and speaker
- Housed within a waterproof, injection-molded, high-strength polymer case
Order Info
Weight: 8 oz. Batteries: two “AAA”, last up to 18 hours.
Certifications
NIOSH rated as intrinsically safe for use in Class I, II, and III, Division 1, Groups A, B, C, D, E, F, and G hazardous locations.
Specifications
AAA | |
Microphone and speaker, push-button on/off | |
0.226 kg | |
Millennium™ and advantage™ 1000 facepiece |
NIOSH | |
8 oz. | |
Hands-free voice amplification for local, short-range communication | |
Voice Amplifier |