MSA™ Comfo™ Cartridges
Particulate, chemical, and combination cartridges for use with Comfo Classic™, Comfo Elite™, Ultra-Twin™ and Ultra Elite™ respirators
Manufacturer: MSA™ 464046
Description
- Low-profile particulate and ShortStack™ combination cartridges feature a patented low-profile design that improves field of vision
- Special adapter also allows use with Advantage™ 200LS, Advantage 420, Advantage 4000 and Advantage 1000 respirators Sparkfoe™ particulate cartridge is suitable for use in applications with sparking hazard
Note: Do not exceed maximum use concentrations established by regulatory standards.
Certifications
All Comfo cartridges are approved under NIOSH 42CFR Part 84.
Specifications
Organic Vapor, Acid Gas Cartridge | |
Yellow | |
N95, R951 Filter | |
Asbestos abatement, painting, sanding, grinding, welding | |
Comfo™ Respirator Cartridge |
NIOSH 42CFR, Part 84 | |
95% | |
10/Pk. | |
Comfo classic™, ultra-twin™, ultra elite twin™ cartridge and duo-twin™ respirator and certain half mask and full face advantage™ respirator |