MSA™ Bump Cap
Protect your workers from minor bumps and lacerations
Manufacturer: MSA™ 10033651
Description
- Size-adjustable; no replaceable parts
- Ideal for workplaces such as meat packing, food processing, pest control, meter reading, and automotive repair
- Perforated sides improves air circulation by allowing cross ventilation
- Interior plastic liner includes foam pad for additional comfort
- Standard version is molded from durable high-density polyethylene and is adjustable from size 6.5 to 8
NOTE: Bump caps do not meet the ANSI Z89.1-1997 standard, and are not designed to protect against impact of falling or glancing objects.
Specifications
Yellow | |
Standard |