MSA™ Bump Cap
Protect your workers from minor bumps and lacerations
Supplier:  MSA™ 10033650
 View more versions of this product
Catalog No. 29-635-1358
Description
Description
  • Size-adjustable; no replaceable parts
  • Ideal for workplaces such as meat packing, food processing, pest control, meter reading, and automotive repair
  • Perforated sides improves air circulation by allowing cross ventilation
  • Interior plastic liner includes foam pad for additional comfort
  • Standard version is molded from durable high-density polyethylene and is adjustable from size 6.5 to 8
NOTE: Bump caps do not meet the ANSI Z89.1-1997 standard, and are not designed to protect against impact of falling or glancing objects.
Specifications
Specifications
Blue
HDPE
Standard
Adjustable