missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MSA™ Assay Probe Tubes
$455.39 - $455.39
Specifications
Type | Probe Tube |
---|---|
For Use With (Application) | Measures concentrations of combustible gases or vapors in air |
Length (English) | 3 ft. |
Length (Metric) | 914 mm |
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
2985176
|
MSA™
11961 |
Brass |
Each for $455.39
|
|
Specifications
Probe Tube | |
Measures concentrations of combustible gases or vapors in air | |
3 ft. | |
914 mm |