MSA™ AirHawk™ SCBA Stock Configurations
Popular configurations
Manufacturer:  MSA™ 10043202
 View more versions of this product
Catalog No. 29-812-141
Description
Description
For detailed specifications, consult general description for Airhawk SCBAs or contact your Fisher Safety Customer Service Representative.
  • SCBA Respirator, 2216
  • Harness: Nylon with chest strap
  • Facepiece: Ultra Elite, medium, Hycar, rubber head harness
  • Cylinder: Aluminum (30 minutes)
Specifications
Specifications
Airhawk SCBA with no case
2216 psig
Medium Hycar™
30 min.
Nylon with chest strap, rubber