MSA™ Advantage™ Cartridges
Lightweight for comfort, but designed for maximum protection
Manufacturer:  MSA™ 815361
 View more versions of this product
Catalog No. 189993765
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
Description
  • Particulate, chemical, and combination cartridges
  • Bayonet-style mounting allows easy installation—cartridges lock into place with a single twist
  • Interchangeable between Advantage 200LS, Advantage 420, Advantage 1000, Advantage 4000 and Advantage 3200 respirators
Order Info
Packaged two cartridges per pack.
Compliance
NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirements; EN 14387. Meets OSHA 29 CFR Part 1920.134.
Specifications
Specifications
Mercury Vapor Cartridge
Bayonet
2/Pk.
Advantage™ 200 ls and 420 half masks and full facepiece respirator (1000, 3000 and 4000 series)
OSHA, NIOSH 42CFR, Part 84, EN 14387, AS NZS 1716, GOST, DSTU
N95, R951 Filter
Asbestos abatement, welding, sanding, grinding, painting, public events, search, rescue
Advantage™ Respirator Cartridge