missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSA™ Advantage™ 420 Half-Mask Respirator
One-piece UniBond over-molded silicone facepiece helps eliminate possible leak paths
$73.81 - $78.55
Specifications
| Material | Silicone |
|---|---|
| For Use With (Application) | Asbestos abatement, painting, sanding, grinding, welding |
| For Use With (Equipment) | Respiratory Systems |
| Latex Free | Yes |
| Catalog Number | Mfr. No. | Model | Includes | Size | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Model | Includes | Size | Price | Quantity | |||
|
19-825-001
|
MSA™
10102183 |
Advantage 420 Respirator Assembly | Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve | Medium |
Each for $78.55
|
|
|||
|
19-825-000
|
MSA™
10102182 |
Advantage 420 Respirator Assembly | Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve | Small |
Each for $74.03
|
|
|||
|
19-825-002
|
MSA™
10102184 |
Advantage 420 Respirator Assembly | Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve | Large |
Each for $73.81
|
|
|||
Description
- Drop-down harness position allows respirator to be removed while retaining head protection
- One-piece ComforTop™ cradle headband eliminates pressure points
- AnthroCurve™ multi-ethnic face seal improves fit-testing among diverse workforce
- Lock-down harness position allows a personal fit without further user adjustments
- No loose or dangling straps to impede operation or vision
- Latex-free; high-quality silicone provides greater durability and product life
- Textured inner facepiece seal minimizes slippage, improves fit
- Bayonet and threaded cartridge options allow use of all MSA cartridges
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
Meets OSHA 29 CFR Part 1920.134
Specifications
| Silicone | |
| Asbestos abatement, painting, sanding, grinding, welding | |
| Respiratory Systems | |
| Yes |