MSA™ Advantage™ 420 Half-Mask Respirator
One-piece UniBond over-molded silicone facepiece helps eliminate possible leak paths
Supplier: MSA™ 10102184
Description
- Drop-down harness position allows respirator to be removed while retaining head protection
- One-piece ComforTop™ cradle headband eliminates pressure points
- AnthroCurve™ multi-ethnic face seal improves fit-testing among diverse workforce
- Lock-down harness position allows a personal fit without further user adjustments
- No loose or dangling straps to impede operation or vision
- Latex-free; high-quality silicone provides greater durability and product life
- Textured inner facepiece seal minimizes slippage, improves fit
- Bayonet and threaded cartridge options allow use of all MSA cartridges
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
Meets OSHA 29 CFR Part 1920.134
Specifications
NIOSH, AS/NZS 1716:2003, 42 CFR 84 | |
Half Mask Respirator | |
Asbestos abatement, painting, sanding, grinding, welding | |
Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve | |
Advantage 420 Respirator Assembly |
Silicone | |
Large | |
Respiratory Systems | |
Yes |