missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MSA™ Advantage™ 3200 Twin-Port Respirator
Fit is based, in part, on facial measurement data from more than 8000 people
$283.78 - $311.98
Specifications
Certifications/Compliance | NIOSH 42CFR84, ANSI Z87.1, EN136 Class 2 |
---|---|
Connection Type | Dual-canister mount (bayonet mount) |
Product Type | Full Face Respirator |
For Use With (Application) | Agriculture, chemical construction, electric utility, mining, nuclear oil and gas, paper and pulp sanding and grinding welding, asbestos, abatement |
Material | Silicone |
Requires: NIOSH-approved Advantage series cartridges and filters.
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-041-906A
|
MSA™
10028996 |
Small |
Each for $283.78
|
|
19-041-906B
|
MSA™
10028995 |
Medium |
Each for $309.82
|
|
19-041-906C
|
MSA™
10028997 |
Large |
Each for $311.98
|
|
Description
- Twin-port 3200 respirator has lower breathing resistance than traditional full-facepiece respirators
- Soft, pliable 3000 Series silicone facemask and rubber head harness don quickly
- Pre-adjustable straps ensure a consistent fit and tighter seal
- Optically correct, wraparound facepiece lens provides a wide, distortion-free field of vision protected by a scratch-resistant lens coating and a fog-reducing standard nosecup
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters. Facepiece lens exceeds the performance requirements of ANSI™ Z87.1.
Specifications
NIOSH 42CFR84, ANSI Z87.1, EN136 Class 2 | |
Full Face Respirator | |
Silicone | |
0.522 kg |
Dual-canister mount (bayonet mount) | |
Agriculture, chemical construction, electric utility, mining, nuclear oil and gas, paper and pulp sanding and grinding welding, asbestos, abatement | |
1.15 lb. |