MSA™ Advantage™ 200 LS Half-Mask Respirators
Half-mask respirators offer maximum protection and high wearing comfort with its patented MultiFlex system. Ideal for applications where workers are exposed to various hazards such as high concentrations of fumes, mists and gases. Available in single or two-piece neckstrap.
Supplier: MSA™ 815700
Alert:
Cartridges and filters sold separately
Requires : Advantage cartridges and filters (see separate listings). Also compatible with Comfo Series cartridges when used with adapter 18-999-1504.
Description
- Comfortable, efficient and economic half mask
- Light, soft pliable thermoplastic facepiece
- Multiflex&;trade; System equalizes pressure on the face-seal area while delivering stability
- Available with single or two-piece neck strap
- Ideal for applications where workers are exposed to high concentrations of fumes, mists and gases
- Customized and pressure-free fit, instantly conforms to the wearer’s unique facial characteristics
- Available in three sizes and has a full range of cartridges and filters to protect against harmful particles and gases
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
EN 140 | |
Blue | |
Half-Mask Respirator | |
Asbestos abatement, painting, sanding, grinding, welding | |
815700 | |
4-point |
NIOSH 42 CFR Part 84 | |
Thermoplastic Elastomer | |
Large | |
Advantage™ Flexi-Filters™ and Advantage™ Cartridges | |
Two-piece |