Learn More
MSA™ Advantage™ 1000 Respirator
Wraparound flexible lens offers excellent fit and comfort with increased vision
$661.31 - $690.63
Specifications
Certifications/Compliance | AS NZS 1716:2003, NIOSH |
---|---|
Color | Black |
Product Type | Full Face Respirator |
For Use With (Equipment) | Msa™ cartridge and filter |
Includes | Speaking diaphragm and nosecup |
Requires: Advantage cartridges and filters (sold separately). Also compatible with Comfo Series cartridges when used with adapter 18-999-1504.
Catalog Number | Mfr. No. | Size | Weight (English) | Weight (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Weight (English) | Weight (Metric) | Price | Quantity | |||
18-999-1507
|
MSA™
805408 |
Medium | 0.8 lb. | 0.363 kg |
Each for $661.31
|
|
18-999-1506
|
MSA™
805420 |
Large | 1.25 lb. | 0.567 kg |
Each for $690.63
|
|
18-999-1508
|
MSA™
805414 |
Small | 1.15 lb. | 0.522 kg |
Each for $666.92
|
|
Description
Low-maintenance, full-face respirator weighs 40% less than conventional full-face respirators. Constructed of exceptionally soft Hycar™ rubber that provides the feel and comfort of silicone but with better permeation resistance.
- Fully elastic, six-point head harness adjusts easily
- Interchangeable Advantage Series cartridges enhance versatility and are compatible with Advantage 200, Advantage 420, Advantage 4000 and Advantage 3200 respirators
- Unique inturned lip seal on facepiece provides an exceptional seal against a wide range of facial contours
- Flexible urethane lens is integrally bonded to the facepiece, eliminating the need for rigid lens retainers
- Includes a speaking diaphragm for clear short-range communications and a nosecup to reduce lens fogging in low temperatures or high humidity
Order Info
Optional lens cover, lens shields, and ESP™ communication system available separately.
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
AS NZS 1716:2003, NIOSH | |
Full Face Respirator | |
Speaking diaphragm and nosecup | |
Urethane |
Black | |
Msa™ cartridge and filter | |
Hycar™ Rubber |