MSA™ Advantage™ 1000 CBA-RCA and Millennium™ CBRN Gas Mask Replacement Canisters
For use with 40mm threaded Millennium CBRN Gas Masks
Manufacturer: MSA™ 817590
Description
- Replacement canisters for CBRN gas masks
- Contain chemical sorbents and a P100 filter to attract, retain and neutralize contaminants
Specifications
NIOSH CBRN | |
Black | |
Gas mask |
Bayonet | |
Canister | |
6/Pk. |