MSA™ Advantage™ 1000 CBA-RCA and Millennium™ CBRN Gas Mask Replacement Canisters
For use with 40mm threaded Millennium CBRN Gas Masks
$111.98 - $581.85
Specifications
Certifications/Compliance | NIOSH CBRN |
---|---|
Product Type | Canister |
For Use With (Equipment) | Gas mask |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
19-130-0457
|
MSA™
10046570 |
1 Ea. |
Each for $111.98
|
|
29812071
|
MSA™
817590 |
6/Pk. |
Pack of 6 Each for $581.85
|
|
Description
- Replacement canisters for CBRN gas masks
- Contain chemical sorbents and a P100 filter to attract, retain and neutralize contaminants
Specifications
NIOSH CBRN | |
Canister | |
Gas mask |