Please login to your online account to display your discounted pricing

MS 245 oxalate, Tocris™

High affinity 5-HT6 antagonist

$251.85 - $1,045.45

Chemical Identifiers

Chemical Name or Material MS 245 oxalate
Assay Percent Range >99%
Molecular Formula C19H22N2O3S·C2H2O4
Formula Weight 448.49
Synonym 5-Methoxy-N, N-dimethyl-1-(phenylsulfonyl)-1H-indole-3-ethanamine oxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

420850

 
r & d systems
RDS420850
50mg Each for $1,045.45

420810

 
r & d systems
RDS420810
10mg Each for $251.85
Description & Specifications

Specifications

Formula Weight 448.49
Quantity 50mg
Chemical Name or Material MS 245 oxalate
Assay Percent Range >99%
Molecular Formula C19H22N2O3S·C2H2O4
CAS 275363-58-1
Synonym 5-Methoxy-N, N-dimethyl-1-(phenylsulfonyl)-1H-indole-3-ethanamine oxalate
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C