Please login to your online account to display your discounted pricing

MRT 10, Tocris™

Smoothened (Smo) antagonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material MRT 10
Assay Percent Range >98%
Molecular Formula C24H23N3O5S
Formula Weight 465.52
Synonym N-[[[3-Benzoylamino)phenyl]amino]thioxomethyl]-3, 4, 5-trimethoxybenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

458510

 
r & d systems
RDS458510
10mg Each for $225.77

458550

 
r & d systems
RDS458550
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 465.52
Quantity 10mg
Chemical Name or Material MRT 10
Assay Percent Range >98%
Recommended Storage Store at +4°C
Molecular Formula C24H23N3O5S
CAS 330829-30-6
Synonym N-[[[3-Benzoylamino)phenyl]amino]thioxomethyl]-3, 4, 5-trimethoxybenzamide
Safety and Handling

Store at +4°C